ContactUsLinkComponent
PREempt* Concentrate, 1 gallon
PREempt® Concentrate Disinfectants, Contec
Catalog #: CA10836-022
Supplier:  Contec
PREempt* Concentrate, 1 gallon
PREempt® Concentrate Disinfectants, Contec
Catalog #: CA10836-022
Supplier:  Contec
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    4 L
  • Description:
    PREempt® Concentrate Disinfectant, Capped
  • Cat. no.:
    CA10836-022
  • Packaging:
    4/case
  • Supplier No.:
    11305
Specifications
About this item
One-step disinfectants cleaner utilizing Accelerated Hydrogen Peroxide®.
  • Concentrate solution is cost effective for daily use in large areas
  • Low residue and compatible with a wide range of materials
  • Odor-free for good operator acceptability
  • Does not require personal protection equipment (PPE) once diluted
  • Not classified as dangerous goods – no special shipping required
PREempt® concentrate is a patented synergistic blend of commonly used ingredients that when combined with low levels of hydrogen peroxide dramatically increase its potency and cleaning performance. AHP combines hydrogen peroxide with surfactants, wetting and chelating agents to create an ideal one-step disinfectant cleaner.
AHP is a unique formulation that has a perfect balance between a fast contact time while maintaining cleaning efficacy.
It requires dilution creating a cost effective solution for cleaning and disinfecting large areas of cleanroom or controlled environments. It leaves minimal residues on surfaces when dry.
For use in laboratories and other cGMP facilities, clean rooms and other critical environments that require cleaning and surface disinfection. This includes workstations, fume hoods, laboratory countertops, equipment and other hard non-porous environmental surfaces.
5 minutes bactericidal, virucidal, fungicidal (1:40)
5 minutes tuberculocidal (1:40)
30-second broad-spectrum sanitizing (1:128)
Customers Who Bought This Also Bought