Versaflo™ S-Series Hoods with Integrated Head Suspension, 3M™
Catalog #: CA89132-116
Supplier: 3M
Specifications
- Description:Hood, M-L
- Cat. no.:CA89132-116
- Supplier No.:S-433L-5
Specifications
About this item
Fully assembled hood provides head, face, neck, and shoulder coverage.
- Integrated comfort suspension has a wide faceseal with soft elastic for improved comfort and reduced airflow noise
- Exhalation panel vents the headcover without the traditional vent hole pattern or exhalation valve
- Visor has an excellent field of view with reduced curvature for decreased reflections and glare
Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or in situations where there is less soiling of the outer fabric. Approved for use with certain 3M™ powered air purifying and supplied air respirator systems.
Certifications: OSHA APF 1000.
Customers Who Bought This Also Bought