VWR® Pipette Carousel Stand
Catalog #: 10827-940
Supplier: VWR International
Specifications
- Description:VWR® Carousel Pipettor Stand
- Cat. no.:10827-940
Specifications
About this item
Rotary pipette stand for up 7 pipettors. Made from PP/PC.
- Pipettes hang vertically to prevent liquids from running inside the shaft
- Rotating head for easy access
- Designed for single and multichannel models
Customers Who Bought This Also Bought