Vacuum Take-off Short Stem Bend Adapter
Catalog #: CA10823-638
Supplier: CHEM SCIENCE INC CA
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78