ContactUsLinkComponent
Antibiotic Assay Disc
Whatman™ Antibiotic Assay Paper, Whatman products (Cytiva)
Catalog #: CA89007-048
Supplier:  Cytiva
CAS Number:  
Antibiotic Assay Disc
Whatman™ Antibiotic Assay Paper, Whatman products (Cytiva)
Catalog #: CA89007-048
Supplier:  Cytiva
CAS Number:  
Specifications
  • Description:
    Antibiotic Assay Discs, 6 mm
  • Supplier No.:
    2017-006
Specifications
About this item
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method. The antibiogram allows rational and selective chemotherapy. The test disks are coated with chemotherapeutic agents, placed on the inoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.
The antibiogram allows rational and selective chemotherapy. The test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated. The size of the inhibition zone is a measure for the effectiveness of the substances.
Customers Who Bought This Also Bought