ContactUsLinkComponent
PH indicator Strips 6.0 to 8.1
Whatman™ pH Indicators with Integral Comparison Strips, Whatman products (Cytiva)
Catalog #: CA70250-026
Supplier:  Cytiva
Whatman™ pH Indicators with Integral Comparison Strips, Whatman products (Cytiva)
Catalog #: CA70250-026
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Dimensions:
    11 x 100 mm
  • Description:
    Integral Comparison Strip, narrow range
  • pH Range:
    6 - 8.1
  • Supplier No.:
    2629-990
Specifications
About this item
Can be used for the rapid determination of pH values has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use
Each test strip has a central segment of indicator dye and, printed alongside, eight or more different color segments marked with corresponding pH values for matching purposes. The pH test value can be read off by direct comparison of the test strip color and the color bars. Excellent for colored solutions, when any changes in color of the paper stock are automatically cancelled out.
Customers Who Bought This Also Bought