Whatman™ Benchkote Surface Protector, Whatman products (Cytiva)
Catalog #: CA52857-007
Supplier: Cytiva
Specifications
- Dimensions:920 mm × 50 m
- Description:Benchkote surface protector
- Cat. no.:CA52857-007
- Type:Roll
- Supplier No.:2300-772
Specifications
About this item
Cytiva's Whatman Benchkote surface protector is an absorbent lab paper that protects surfaces against hazardous spills. After use, the bench protector sheet is incinerated or disposed of according to local regulations.
- Benchkote surface protector is strong and tear resistant when wet or dry
- Smooth white surface can be written on with ink or pencil and lies flat
- Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
- Can be incinerated after use: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
Benchkote surface protector minimizes mess and contamination during spills. Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Customers Who Bought This Also Bought