Whatman™ Grade GF/C Glass Microfiber Filters, Binder Free
Catalog #: CA28497-641
Supplier: Cytiva
Specifications
- Particle retention:1.2 µm
- Description:Grade GF/C glass microfiber filter, 24 mm circle
- Material:Borosilicate glass
- Size:100 Pieces
- Basis weight:53 g/m²
- Max. operating temperature:550 °C
- Thickness:260 µm
- Filtration speed:Medium to fast, 100 s (Herzberg)
- Water flow rate:105 ml/min
- Airflow rate:6.7 s/100 ml/in²
- Binder:Binder-free
- Cat. no.:CA28497-641
- Grade:GF/C
- Supplier No.:1822-024
Specifications
About this item
These borosilicate glass filters are widely used as total suspended solids filters for applications testing both potable water and waste water. GF/C can also function as a filter for liquid scintillation counting, cell harvesting, industrial waste analysis and binding assays that require high loading capacity.
- Fine particle retention
- Good flow rate and loading capacity
- Binder free
Customers Who Bought This Also Bought