Order Entry
ContactUsLinkComponent
VWR* Qualitative Filter Paper 15cm
VWR® Qualitative Crepe Fluted Circles, Folded, Grade 315
Catalog #: CA28331-048
Supplier:  VWR International
VWR* Qualitative Filter Paper 15cm
VWR® Qualitative Crepe Fluted Circles, Folded, Grade 315
Catalog #: CA28331-048
Supplier:  VWR International

Specifications

  • Ø:
    15.0 cm
  • Particle retention:
    25 µm
  • Flow rate:
    Fast
  • Porosity:
    Coarse

Specifications

About this item

Crepe fluted circles are ideal for gelatinous and coarse precipitates.

  • Coarse porosity
  • Fast flow rate
  • Assorted sizes
Customers Who Bought This Also Bought