VWR® Qualitative Crepe Filter Paper, Grade 417
Catalog #: CA28313-160
Supplier: VWR International
Specifications
- Ø:25.0 cm
- Particle retention:40 µm
- Porosity:Coarse
Specifications
About this item
Good for rapid filtrations where fine particle retention is not required.
- Coarse porosity
- Fast flow rate
- Assorted sizes
Customers Who Bought This Also Bought