Order Entry
ContactUsLinkComponent
VWR* Filter Paper Grade 413 12.5cm
VWR® Qualitative Filter Paper, Grade 413
Catalog #: CA28310-106
Supplier:  VWR International
VWR® Qualitative Filter Paper, Grade 413
Catalog #: CA28310-106
Supplier:  VWR International

Specifications

  • Ø:
    125 mm
  • Particle retention:
    5 µm
  • Flow rate:
    Medium
  • Porosity:
    Medium

Specifications

About this item

Suitable for filtration of aluminum phosphate, barium carbonate, lead chromate, and more.

  • For routine lab work

Suitable for student requirements in general chemistry.

Customers Who Bought This Also Bought