Order Entry
ContactUsLinkComponent
VWR* Filter Paper Grade 474 12.5cm
VWR® Quantitative Filter Paper, Grade 474
Catalog #: CA28306-380
Supplier:  VWR International
VWR* Filter Paper Grade 474 12.5cm
VWR® Quantitative Filter Paper, Grade 474
Catalog #: CA28306-380
Supplier:  VWR International

Specifications

  • Ø:
    12.5 cm
  • Particle retention:
    5–10 µm
  • Flow rate:
    Medium Slow
  • Porosity:
    Medium Fine

Specifications

About this item

Uses include filtration of aluminum phosphate, zinc ammonium phosphate, and silver chromate.

For general quantitative analyses.

Customers Who Bought This Also Bought