Order Entry
ContactUsLinkComponent
VWR* Filter Paper Grade 454 12.5cm
VWR® Quantitative Filter Paper, Grade 454
Catalog #: CA28306-131
Supplier:  VWR International
VWR* Filter Paper Grade 454 12.5cm
VWR® Quantitative Filter Paper, Grade 454
Catalog #: CA28306-131
Supplier:  VWR International

Specifications

  • Ø:
    12.5 cm
  • Particle retention:
    10 µm
  • Flow rate:
    Medium Fast
  • Porosity:
    Medium

Specifications

About this item

Use for retention of zirconium hydroxide, aluminum hydroxide, cobalt sulfide, and other coarse or gelatinous precipitates.

Customers Who Bought This Also Bought