Order Entry
ContactUsLinkComponent
Cellulose Blotting Paper GB003 15X15cm
Whatman™ Cellulose Blotting Papers, Grade GB003
Catalog #: CA28153-293
Supplier:  Cytiva
Cellulose Blotting Paper GB003 15X15cm
Whatman™ Cellulose Blotting Papers, Grade GB003
Catalog #: CA28153-293
Supplier:  Cytiva

Specifications

  • Dimensions:
    15×15 cm
  • Description:
    Grade GB003 Blotting paper
  • Thickness:
    0.8 mm
  • Grade:
    GB003
  • Supplier No.:
    10427810

Specifications

About this item

This general purpose blotting paper is made from pure raw materials with a high absorbency and is used as a membrane gel support. Due to its thickness (0.8 mm), it is recommended for the lysis/denaturation of colony or plaque lifts and Western blots.

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives ensures that no contamination will occur during the transfer steps
  • Manufactured and tested specifically for chromatographic and blotting techniques ensuring the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting
  • Industry standard for blotting procedures
  • Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes allows 'out-of-the-box' usage and eliminates sheet-to-sheet variation

A thick paper recommended for the lysis/denaturation of colony or plaque lifts and Western blots.

Customers Who Bought This Also Bought