Order Entry
ContactUsLinkComponent
Vacuguard Disc Filter 50mm
Whatman™ VACU-GUARD Disk Filters, Whatman products (Cytiva)
Catalog #: CA28137-858
Supplier:  Cytiva
Vacuguard Disc Filter 50mm
Whatman™ VACU-GUARD Disk Filters, Whatman products (Cytiva)
Catalog #: CA28137-858
Supplier:  Cytiva

Specifications

  • Ø:
    50 mm
  • Housing material:
    Polypropylene (PP)
  • Pore size:
    0.45 mm
  • Description:
    VACU-GUARD 50 mm Disc, PTFE Polytetrafluoroethylene
  • Filtration Area:
    16 cm²
  • Supplier No.:
    6722-5000
  • Retention:
    Aqueous solutions: up to 0.9 bar (14 psi)
    Particulates in air: 0.1 µm 99.99%
  • Max. operating pressure:
    1 bar (15 psi)
  • Application:
    Vacuum pump protection.
    Activated carbon removes organic vapors and radioactive particles.
    Molecular sieve for use with organic and alkaline air streams.
    Desiccant for use with high velocity acidic air streams.
    Removes a potential health hazard from the w

Specifications

About this item

Whatman VACU-GUARD disk filters are ideal for protecting vacuum pump systems from aqueous aerosols and water intrusion.

  • Designed for in-line use with stepped barb connections
  • Membrane retains 99.99% of airbone particles greater than 0.1 µm
  • Features hydrophobic polytetrafluoroethylene (PTFE) membrane

Available in 50 and 60 mm formats.

Ordering information: All materials pass USP Class VI test for plastics.
Customers Who Bought This Also Bought