EDTA disodium salt dihydrate 99.0-101.0% ACS, VWR Chemicals BDH®
Catalog #: BDH4616-500G
Supplier: VWR International
CAS Number: 6381-92-6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
- Size:500 g
- Supplier No.:BDHVBDH4616-500G
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- pH (25°C; 5 %):4.0 - 6.0
- Heavy metals (as Pb):Max. 0.005 %
- Insoluble matter:Max. 0.005 %
- Nitrilotriacetic acid:Max. 0.1 %
- Fe (Iron):Max. 0.01 %
- VWR Chemicals BDH®, EDTA disodium salt dihydrate 99.0-101.0% ACS:
Specification Test Results
About this item
Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate,Kestranal® 2S dihydrate,Disodium dihydrogen ethylenediaminetetraacetate dihydrate,Ethylenedinitrilotetra-acetic acid disodium salt dihydrate,Titriplex III,Ethylenediaminetetra-acetic acid disodium salt dihydrate,Kestranal® 2S 2 H2O,Edetate disodium dihydrate
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt:255 °C
MDL Number:MFCD00150037
CAS Number:6381-92-6
:EDTA disodium salt dihydrate
:Storage Temperature: Ambient
:Length
:Width
:Height
:Diameter
:Thickness
:99,0-101,0
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 255 °C
:Flash Pt:
:Flash Pt:
:M.W. 372.24 g/mol
Merck Index:13,03543
Storage Temperature:Ambient
EINECS:205-358-3
Customers Who Bought This Also Bought