Anti-NEFH Mouse Monoclonal Antibody [clone: SMI 32]
Catalog #: CA10816-352
Supplier: Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Neurofilament, Heavy Polypeptide
- Clonality:Monoclonal
- Clone:SMI 32
- Gel shift:Yes
- Gene ID:4744
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mammals,Mouse
- Antigen symbol:NEFH
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µL
- Western blot:Yes
- Form:Phosphate-buffered solution.
- Antigen synonyms:NFH
- Storage temperature:The antibody solution should be stored undiluted between 2°C and 8°C.
- Concentration:1 mg/ml
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography.
- Cat. no.:CA10816-352
- Supplier No.:801701
Specifications
About this item
Type: Primary
Antigen: NEFH
Clonality: Monoclonal
Clone: SMI 32
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammalian, Mouse
Customers Who Bought This Also Bought