To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG1, k Isotype Ctrl
- Clonality:Monoclonal
- Clone:MOPC-21
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:IgG1, k Isotype Ctrl
- Conjugation:Brilliant Violet® 785
- Size:100 Tests
- Epitope:kappa-chain
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Purification:The immunoglobulin was purified by affinity chromatography and conjugated with Brilliant Violet 785™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 785™ and unconjugated immunoglobulin.
- Cat. no.:CA10761-080
- Supplier No.:400170
Specifications
About this item
Brilliant Violet 785™ Mouse IgG1, κ Isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgG1, k Isotype Ctrl
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse