Seal, Constant Level Device, Fi-Streem (Old Style) 2 Liter Replacement Part
Catalog #: CA10809-602
Supplier: LABSTRONG CORPORATION CA
Seal, Constant Level Device, Fi-Streem (Old Style) 2 Liter Replacement Part
Catalog #: CA10809-602
Supplier: LABSTRONG CORPORATION CA
Customers Who Bought This Also Bought
Hydrogen peroxide 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Supplier: VWR International
$568.42
American White Cross First Aid® Herbie® the Dinosaur Bandages, DUKAL™ Corporation
Supplier: Dukal
$13.15 to $157.72
Rapid Response™ COVID-19 Antigen Rapid Test Cassettes - At Home, BTNX
Supplier: BTNX INC CA
$21.01 to $5,041.08
Silver nitrate ≥99.995% (metals basis) Ag ≥ 63%, Premion®
Supplier: Thermo Scientific Chemicals
$501.00
Pyruvaldehyde (methylglyoxal) 35 - 45% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
$49.00