Reinforced Pressure Tubing
Catalog #: CA10809-396
Supplier: LABSTRONG CORPORATION CA
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Argos Technologies
$62.31
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50