Preformed Nylon pipe (solv/dump valve), Replacement part for Fi-Streem Water Purification Stills
Catalog #: CA10809-348
Supplier: LABSTRONG CORPORATION CA
Preformed Nylon pipe (solv/dump valve), Replacement part for Fi-Streem Water Purification Stills
Catalog #: CA10809-348
Supplier: LABSTRONG CORPORATION CA
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Copper(II) sulfate pentahydrate 98.0-102.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$105.35 to $893.56
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$483.15 to $1,605.80
Hydrochloric acid 36.5 - 38.0% NF, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$564.79