Straight Nipple Cartridge
Catalog #: CA10809-206
Supplier: LABSTRONG CORPORATION CA
Customers Who Bought This Also Bought
Bromophenol blue 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$91.88 to $955.56
Palladium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®, VWR®
Supplier: VWR International
$468.98
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
$90.42 to $94.61