Order Entry
ContactUsLinkComponent
VWR* Animal Ear Punch without tags thumb style, Length: 2.00 mm
VWR® Animal Ear Punches
Catalog #: 10806-286
Supplier:  VWR International
VWR* Animal Ear Punch without tags thumb style, Length: 2.00 mm
VWR® Animal Ear Punches
Catalog #: 10806-286
Supplier:  VWR International

Specifications

  • Ø:
    2 mm Hole Size
  • Length:
    5 cm (2")
  • Description:
    Animal Ear Punch, Thumb-Style, without Tags
  • Material:
    Surgical Grade Stainless Steel
  • Cat. no.:
    10806-286

Specifications

About this item

VWR® Animal Ear Punches are made from corrosion-resistant ASTM (899-94) Series 400 stainless steel.

  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

These ear punches are available in multiple designs, including thumb-style, plier-style, or scissor-style punches. Both the plier-style and scissor-style punches feature a longer handle for improved grip and handling.

Customers Who Bought This Also Bought