Order Entry
ContactUsLinkComponent
Mycoplasma Removal Agent
Mycoplasma Removal Agent (MRA), MP Biomedical
Catalog #: CAIC3050044
Supplier:  MP Biomedicals
CAS Number:  
Mycoplasma Removal Agent
Mycoplasma Removal Agent (MRA), MP Biomedical
Catalog #: CAIC3050044
Supplier:  MP Biomedicals
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mycoplasma Removal Agent
  • Size:
    5 ml
  • Sterility:
    Sterile
  • Concentration:
    50 µg/ml (Lit.)
  • Cat. no.:
    CAIC3050044

Specifications

About this item

Mycoplasma removal agent (MRA) is a highly effective solution for the serious problem of mycoplasma contamination in cell cultures.

  • Eliminates multiple mycoplasma species within one week using the lowest dosage
  • Prevents recontamination of the culture at 0.1 µg/ml
  • Maintains cell viability
  • Compatible with most mammalian cell lines
  • Sterility and low cytotoxicity

Mycoplasma Removal Agent (MRA) from MP offers a highly effective solution for the serious problem of mycoplasma contamination in cell cultures. MRA is a derivative of the quinoline family of antibiotics and eliminates mycoplasma infection (recommended use at a concentration of 0.5 µg/ml) by inhibiting mycoplasma DNA gyrase. MRA is supplied as a ready-to-use solution and each vial can decontaminate up to 25 cell cultures. MRA has been cited by the European collection of animal cell cultures (porton down, U.K.) as being superior to all other current products for the eradication of mycoplasma. Furthermore, cell toxicity is low following treatment at the recommended concentration, as is regrowth of mycoplasma after cellular transfer.

Mycoplasma removal agent (MRA) eliminates mycoplasma contamination from cell cultures. The cytotoxicity of MRA is low and cell toxicity is rare when used at the recommended concentration. For specific function of any cell, however, it is recommended that the retention of desired cellular characteristics be confirmed after treatment.

Concentration may be raised up to 1 µg/ml only when the recommended concentration is ineffective in removing the mycoplasma. For preventative purposes, MRA can be used at 0.1 µg/ml.

Ordering information: Supplied in a 5 ml amber glass bottle. Ready-to-use solution decontaminates up to twenty-five cell cultures.
Caution: Store at room temperature (15...30 °C), protect from light.
Customers Who Bought This Also Bought