Screw Caps, MicroSolv™
Catalog #: 10804-658
Supplier: MicroSolv Technology Corp
Specifications
- Cap Size:9-425 mm
- Cap Type:Screw cap
- Septum:White Silicone Rubber / White PTFE Pre-Slit, fitted
- Material:PP
- Cat. no.:10804-658
- Cap Color:Blue
- Supplier No.:9502S-30M-B