Vial
Catalog #: 10804-196
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Micromesh™ Biopsy Cassettes in Quickload™ Sleeves, Simport Scientific
Supplier: Simport Scientific
942,84 $
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
169,74 $ to 169,74 $
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Supplier: Thermo Scientific Chemicals
56,00 $
Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
169,41 $ to 781,78 $
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
186,99 $
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
120,05 $ to 125,45 $