Specific Heat Demonstration Set
Catalog #: 470218-914
Supplier: VWR International
Specifications
- Description:Wax Paper Replacement, Pack of 2
- Cat. no.:470218-914
- Supplier No.:PH20202S
Specifications
About this item
Visually illustrate the difference in specific heat.
- Illustrate the different heat capacities of metal
A dramatic way to illustrate the different heat capacities of metal, this slide features a slanted panel with four grooves to hold four different metals. Simply place a sheet of bee's wax in the front of the plane and submerge the metal samples in hot water. When the metals are placed on the grooves simultaneously they will melt and sink through to different levels based on their specific heat. Includes four metals cylinders: aluminum, copper, lead, and tin; support stand, two pieces of bees wax, and instructions.
Customers Who Bought This Also Bought