ContactUsLinkComponent
Test strip
Urine Test Strips, Germaine Laboratories
Catalog #: 10024-358
CAS Number:  
Test strip
Urine Test Strips, Germaine Laboratories
Catalog #: 10024-358
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    AimStrip® 10-SG Tests for Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, and Leukocytes in urine
  • Cat. no.:
    10024-358
  • Supplier No.:
    51100
  • No. of Tests:
    100
Specifications
About this item
Reagent strips for the qualitative and semi-quantitative detection of various analytes in urine, up to 10
The MicroAlbumin 2-1 test detects Microalbumin in concentration levels as low as 10 mg/L and Creatinine at 100 mg/L (MicroAlbumin CPT Code 82044 QW, Creatinine CPT Code 82570 QW).
A color chart is provided on each bottle to easily read results.
Certifications: CLIA waived.
Ordering information: AimStrip® 10-SG and AimStrip® US-4OB tests are compatible with the Aimstrip® Urine Analyzer.
Customers Who Bought This Also Bought