ContactUsLinkComponent
Tip
Conductive Liquid Handling (LiHa) Disposable Tips, Tecan
Catalog #: CA10024-080
Supplier:  TECAN US INC. CA
Conductive Liquid Handling (LiHa) Disposable Tips, Tecan
Catalog #: CA10024-080
Supplier:  TECAN US INC. CA
Specifications
  • Volume:
    1000 µl
  • Filter:
    +
  • Tip Style:
    Tecan pure disposable tips, filtered
  • Supplier No.:
    30057817
Specifications
About this item
These disposable tips are carefully designed, produced and validated to ensure reliable performance using Liquid Handling (LiHa) Arms and Flexible Channel (FCA) arms on Tecan platforms.
  • Available in a variety of tip volumes, with or without filters
  • Made of conductive polypropylene
  • Allow capacitive liquid level detection
  • Full product traceability
Tecan standard purity tips are supplied in space-saving cardboard packages and are used in applications such as ELISA, sample handling and biochemical screening.
Tecan pure purity tips are packaged in blister packs, and each lot of tips is tested and certified free from human DNA, DNase, RNase and PCR inhibitors by an external accredited laboratory and are used in applications such as genomics research, forensics and molecular diagnostics.
Tecan sterile purity tips are tested on a lot-by-lot basis in an external accredited laboratory, and are free from human DNA, DNase, RNase and endotoxins. These are certified to be sterile, with a guaranteed Sterility Assurance Level (SAL) of 10⁻⁶ and are supplied packaged in ANSI/SLAS-format boxes and are used in applications such as cell biology, microbiology and cell-based screening.
Delivery: Each tray of Tecan disposable tips is supplied with a Certificate of Conformity (CoC).
Customers Who Bought This Also Bought