Specifications
- Volume:50 µl
- Tip Color:Clear
- Cat. no.:CA10024-060
- Filter:+
- Sterile:+
- Tip Style:Single stack, ANSI/SLAS
- For Use With:MCA 96, MCA 384
- Supplier No.:30038608
Specifications
About this item
Tecan disposable tip portfolio offers a variety of pipetting solutions to complement the MultiChannel Arm 96 for Freedom EVO® and Fluent®* liquid handling workstations.
- Compatible with MultiChannel Arm 96, MultiChannel Arm 384, Multiple Channel Arm 384
- Designed, manufactured, tested and certified to guarantee reliable quality, helping you to achieve optimal results
- Color coded tray colors for easy tip volume identification
- Optimized workstation efficiency and higher flexibility on the deck
Environmental and logistical benefits: The nested tip design simplifies storage, lowers transport costs and produces less waste by reducing storage space requirements by 60%, and using less packaging materials compared with non-nested formats.
Quality and purity: All MCA 96 disposable tips are manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results. Two purity levels are available to suit the needs of your application: Tecan Pure and sterile.
Tecan sterile pipette tips are guaranteed with a sterility assurance level (SAL) of 10-6 all the way, from production, to delivery and finally usage at the customer.
Each Tecan tip box has robust packaging with a shrink wrap and sterile bag to ensure sterility is maintained without any risk of contaminants for the entire shelf life of the product.