Chiller
Catalog #: 10023-986
Supplier: Buchi
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Supplier: Ricca Chemical
$670.90
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Argos Technologies
$62.31
SGE Syringes, General Purpose Manual Syringe, PTFE Tipped Plunger, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
$219.34
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
$186.99
Fused Quartz Microscope Slides and Coverslips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$45.79