ContactUsLinkComponent
Pipet Controller, Ultra, w/Universal Power Supply, 3 0.2um F
Corning® Stripettor™ Ultra Pipette Controller, Corning
Catalog #: 10023-900
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Pipet Controller, Ultra, w/Universal Power Supply, 3 0.2um F
Corning® Stripettor™ Ultra Pipette Controller, Corning
Catalog #: 10023-900
Supplier:  Corning
   Low Manufacturing Impact
Product available on GSA Advantage®
Specifications
  • Description:
    Stripettor™ Ultra Pipette Controller
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Warsaw
  • Cat. no.:
    10023-900
  • Supplier No.:
    4099
Specifications
About this item
This light, ergonomic, motorised pipette controller is designed for use with glass and plastic serological pipettes from 0.5 to 100 ml.
  • Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities
  • Aspirating and dispensing speed is controlled through the finger triggers
  • When fully charged, the batteries enable up to 8 hours of continuous use
  • Large LCD display clearly indicates battery status, pipetting mode, and speed
  • Colored nose pieces for user or application identification
Delivery: Supplied with universal power supply, three 0.2 µm filters, 3 colored nose pieces and a wall hanger. Four samples of each Shorty Stripette serological pipette are also included.
Customers Who Bought This Also Bought