To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Rabbit IgG
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Rabbit
- Antigen symbol:IgG
- Conjugation:CF™ 594
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:50 uL
- Western blot:Yes
- Epitope:Heavy and Light Chain
- Cross adsorption:No
- Form:Liquid
- Antibody fragment:F(ab')2
- Storage buffer:PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide
- Storage temperature:Store at –20 °C.
- Concentration:2 mg/mL
- Purification:Affinity purified
- Supplier No.:20153-1
Specifications
About this item
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™594 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Deep-red fluorescent CF™594 (Ex/Em 593/614 nm) is spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugate, and has excellent photostability.
Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit