Order Entry
ContactUsLinkComponent
E.Z.N.A. Plant DNA DS Kit, 50 prepations
E.Z.N.A.® Plant DNA DS Kit, Omega
Catalog #: CA10017-720
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Plant DNA DS Kit, Omega
Catalog #: CA10017-720
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    E.Z.N.A.® Plant DNA DS Kit
  • Size:
    50 Preps
  • Description:
    E.Z.N.A.® Plant DNA DS Kit
  • Cat. no.:
    CA10017-720
  • Supplier No.:
    D2411-01

Specifications

About this item

The E-Z 96 Plant DNA DS Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a lower DNA content.

  • Up to 50 mg wet tissue can be processed in less than one hour
  • Increase yields and provide high-quality DNA
  • Eliminates the need for chloroform extractions

The E-Z 96 Plant DNA DS Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols, or those having a lower DNA content. Up to 50 mg wet tissue can be processed in less than one hour. The system combines the reversible nucleic acid-binding properties of the HiBind® matrix with the speed and versatility of spin column technology to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates.  Purified DNA is suitable for PCR, restriction digestion, and hybridization applications. This procedure relies on the well-established properties of the cationic detergent, cetyltrimethyl ammonium bromide (CTAB), in conjunction with the unique binding system to increase yields and provide high-quality DNA.  The system eliminates the need for chloroform extractions traditionally associated with CTAB-based lysis methods. Samples are homogenized and lysed in a high salt buffer containing CTAB, binding conditions are adjusted, and DNA is purified using a E-Z 96 DNA Plate. Salts, proteins, and other contaminants are removed to yield high-quality genomic DNA suitable for downstream applications such as endonuclease digestion, thermal cycle amplification, and hybridization applications.

Customers Who Bought This Also Bought