To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plate
- Description:Columbia CNA Agar
- L×W:15 x 100 mm
- Size:19 mL
- Cat. no.:CA10753-962
- Supplier No.:GA50
Specifications
About this item
Columbia CNA (Colistin and Nalidixic Acid) agar with 5% sheep blood, for the selective isolation of Gram- positive bacteria, RSR, reduced stacking ring for automated inoculating devices.
Recommended for use as a selective growth medium for the isolation and differentiation of Gram- positive cocci from clinical and non-clinical specimens with mixed flora.