Order Entry
ContactUsLinkComponent
Triglyceride M
L-Type Triglyceride M Assay, Wako
Catalog #: CA10752-448
Supplier:  Wako Life Sciences
CAS Number:  
L-Type Triglyceride M Assay, Wako
Catalog #: CA10752-448
Supplier:  Wako Life Sciences
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Triglyceride M Enzyme Color B (L-Type)
  • Size:
    4 x 13 mL
  • Storage Temperature:
    +2…+10 °C
  • Supplier No.:
    461-09891

Specifications

About this item

L-Type triglyceride M is an in vitro assay for the quantification determination of triglyceride in serum or plasma.

  • Designed for use with Wako’s assays
  • Determine points of reference
  • Enzymatic method

The Wako Multi-Calibrator Lipids is designed to be used with Wako’s assays to determine the points of reference that are used in the determination of Triglycerides in human serum.

Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought