Order Entry
ContactUsLinkComponent
Basin, 25ml, Sterile, DNA-RNAse and Pyrogen Free, PK5
Pipetting Reservoirs, Argos Technologies
Catalog #: 10015-234
Supplier:  Argos Technologies
Basin, 25ml, Sterile, DNA-RNAse and Pyrogen Free, PK5
Pipetting Reservoirs, Argos Technologies
Catalog #: 10015-234
Supplier:  Argos Technologies

Specifications

  • Sterility:
    Sterile
  • Description:
    Pipetting Reservoirs, 25 mL
  • Cat. no.:
    10015-234
  • Packaging:
    5/Pack
  • Supplier No.:
    B3125-100

Specifications

About this item

These reagent reservoirs are created to work with 8- and 12-channel pipettes

The thick-wall construction of each basin provides a noticeable difference in product and performance. Low cross bars on each end of the basin give the product much more rigidity and durability than competitive offerings.

The reservoirs were developed for ease of use. They feature pour spouts in every corner, and one larger spout on each reservoir to facilitate spill-free pouring. In addition, the individually wrapped and 5-pack versions are packaged in easy-open “Snap & Tear” bags. No need for scissors, or concerns about damaging the basins as the bags are opened.

All three sizes are available in either sterile or nonsterile formats. Each features graduations on the inner walls. Packaging options include bulk (100 pieces), individually packaged, or in bags of 5. The reservoirs are gamma-sterilized and certified RNase-free, DNase-free, and pyrogen-free.

Reservoirs are purposefully designed with angled sides and a narrow V-shaped bottom to ensure that every last drop of costly reagents can be easily aspirated. White polystyrene basins are offered in 25, 50, and 100mL volumes.

Customers Who Bought This Also Bought