UV Crosslinker with Adjustable Height, Boekel Scientific
Catalog #: 10014-738
Supplier: Boekel
CAS Number:
Specifications
- Description:UV Crosslinker AH (Adjustable Height)
- Power:80 W
- Temperature range:Ambient 16...28 °C
- Energy:9,999,900 µJ in 100 µJ increments
- Ext. W×D×H:151/8×143/8×101/4"
- Time:999 m 59 s in 1 s increments
- Cat. no.:10014-738
- Electrical:120 VAC, 50/60 Hz
- Supplier No.:234100
Specifications
About this item
The UV crosslinker is designed for crosslinking of DNA/RNA to cellulose membrane lattice when conducting Northern and Southern Blot experiments.
- Calibrated for 254 nm shortwave, 30 nm and 368 nm bulbs
- Energy mode or Time mode of operation enabled
- Energy efficient electronic ballasts
- Open door safety mechanism
- Large UV protective window
It has an adjustable height feature and includes five 254 nm shortwave bulbs.
The adjustable height feature provides a more accurate reading by keeping the samples on the floor of the chamber, at the same distance from the bulbs as the UV sensor. There is no need to raise up the samples to bulbs, simply adjust the bulbs.
Certifications: UL/CSA/CE Certified.
Customers Who Bought This Also Bought