Order Entry
ContactUsLinkComponent
DNTP Set
dNTP Set, Biotium
Catalog #: 10001-638
Supplier:  Biotium
CAS Number:  
DNTP Set
dNTP Set, Biotium
Catalog #: 10001-638
Supplier:  Biotium
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    dNTP set, 100 mm each
  • Size:
    1 set - 250 µl each
  • Cat. no.:
    10001-638
  • Supplier No.:
    40052

Specifications

About this item

Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mm.

The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.

Customers Who Bought This Also Bought