To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:AlbiQuick™, for Candida Albicans Differentiation, Five Minute Test
- Size:25 tests per kit
- Cat. no.:CA10001-514
- Supplier No.:Z121
Specifications
About this item
The new Hardy AlbiQuick™ utilizes two enzyme color reactions to produce an accurate result that’s easy to read out.
- Test for Candida albicans in only five minutes
- Performed on a convenient card with two reaction wells
- Eliminates lengthy procedures and tedious microscopy
Now you can test for Candida albicans in only five minutes! The new Hardy AlbiQuick™ utilizes two enzyme color reactions to produce an accurate result that’s easy to read out
Requires the use of a handheld 366nm ultra violet lamp.
The old germ tube method of C. albicans identification requires several hours of incubation and tedious microscopy to produce a result. The AlbiQuick™ eliminates the need for this and other lengthy procedures.
A recent study, which compared the above NAG and PRO methods to the traditional germ tube test, showed a sensitivity of 99% and a specificity of 99-100%.