Order Entry
ContactUsLinkComponent
1M TRIETHYLAMMONIUM BICARBONATE
Pierce Triethylammonium Bicarbonate, Thermo Scientific
Catalog #: CAPI90114
Supplier:  Thermo Scientific
CAS Number:  
1M TRIETHYLAMMONIUM BICARBONATE
Pierce Triethylammonium Bicarbonate, Thermo Scientific
Catalog #: CAPI90114
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Triethylammonium Bicarbonate (TEAB), 1M
  • Size:
    50 mL
  • Cat. no.:
    CAPI90114
  • Supplier No.:
    90114

Specifications

About this item

Thermo Scientific Triethylamonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments.

  • Versatile and volatile solvent
  • Easily removed by simple lyophilization

Buffer for use in ion-exchange chromatography and electrophoresis.

Ordering information: Ordering Information: Store protected from direct sunlight in a dry, cool, and well-ventilated area.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought