Order Entry
ContactUsLinkComponent
B-PER COMPLETE 500ML
B-PER™ Pierce™ Complete Bacterial Protein Extraction Reagent, Thermo Scientific
Catalog #: CAPI89822
Supplier:  Thermo Scientific
CAS Number:  
B-PER COMPLETE 500ML
B-PER™ Pierce™ Complete Bacterial Protein Extraction Reagent, Thermo Scientific
Catalog #: CAPI89822
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    B-PER™ Complete bacterial protein extraction reagent
  • Size:
    500 ml
  • Storage conditions:
    Store at 4˚C
  • Supplier No.:
    89822

Specifications

About this item

B-PER™ Complete Bacterial Protein Extraction Reagent is an all-in-one formulation of non ionic detergent and enzymes that enables mild extraction of proteins from both gram-negative and gram-positive bacteria without the need for mechanical disruption. B-PER Complete Reagent combines a universal nuclease and lysozyme to improve cell lysis and reduce viscosity, and may be used for soluble protein extraction and inclusion body purification from bacterial cell lysates.

  • Fast and simple - just add B-PER Reagent to a bacterial pellet, incubate with mixing for 10 to 15 minutes and recover soluble proteins after pelleting the cell debris
  • Convenient - contains lysozyme and a universal nuclease in a single formulation with 4°C storage
  • Excellent yields - recover recombinant proteins from bacterial lysates or purify inclusion bodies to near-homogeneous levels
  • Suitable for any scale of protein extraction
  • Completely compatible with addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (eg. GST, 6xHis) and other applications

B-PER™ Complete Bacterial Protein Extraction Reagents are compatible with downstream applications, such as affinity chromatography (eg. immobilised metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification. Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought