Order Entry
ContactUsLinkComponent
APS MICROSCOPE SLIDE CS1000 YELLOW
APS™ Adhesion Microscope Slides, Matsunami Glass
Catalog #: 10000-146
Supplier:  Matsunami Glass
APS MICROSCOPE SLIDE CS1000 YELLOW
APS™ Adhesion Microscope Slides, Matsunami Glass
Catalog #: 10000-146
Supplier:  Matsunami Glass

Specifications

  • Color:
    Yellow
  • Description:
    Aps™ adhesion slides, with clipped corners
  • L×W:
    75×25 mm (3×1")
  • Thickness:
    1.0 mm
  • Cat. no.:
    10000-146
  • Supplier No.:
    SUAPS12

Specifications

About this item

The Matsunami APS™ slide is treated to provide a strong permanent charge to the slide surface

Matsunami microscope slides are manufactured in Japan using optically clear, exceptionally flat, low fluorescence glass. Slides are available with 90° or clipped corners.

Excellent for routine histology, IHC, and cytology preparations. Optimized for use with thermal printers.

Ordering information: Slides are packaged 100 per box, 10 boxes per case (1000 slides).
Customers Who Bought This Also Bought