Cap
Catalog #: 10803-668
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
$729.52
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44