Cap
Catalog #: 10803-652
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$214.30 to $2,349.04
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$194.14 to $694.12
Pierce™ Coomassie (Bradford) Protein Assay Kits, Thermo Scientific
Supplier: Thermo Scientific
$354.05