Order Entry
ContactUsLinkComponent
FID Jet
Capillary Dedicated FID Replacement Jet for Agilent 5890/6850/6890/7890 GCs, Restek
Catalog #: 10803-252
Supplier:  Restek
FID Jet
Capillary Dedicated FID Replacement Jet for Agilent 5890/6850/6890/7890 GCs, Restek
Catalog #: 10803-252
Supplier:  Restek

Specifications

  • Length:
    43 mm
  • Description:
    Standard, 0.011" ID Tip
  • Cat. no.:
    10803-252
  • For Use With:
    for Agilent 5890/6850/6890/7890 GCs
  • Supplier No.:
    21682

Specifications

About this item

Available in standard or Siltek®-treated high-performance versions.

  • Threads specially coated for easy installation and removal
  • Special processing ensures the highest degree of cleanliness
  • Meets or exceeds original manufacturer's performance

For use with Agilent 5890/6850/6890 GCs. Can be Siltek® treated to be extremely inert, for use with active compounds.

Customers Who Bought This Also Bought