Order Entry
ContactUsLinkComponent
Tube
Amber Plain Disposable Borosilicate Glass Tubes, Kimble Chase
Catalog #: 10799-210
Tube
Amber Plain Disposable Borosilicate Glass Tubes, Kimble Chase
Catalog #: 10799-210

Specifications

  • Length:
    100 mm
  • O.D.:
    16 mm
  • Volume:
    15 ml (overflow capacity)
  • Supplier No.:
    73500-16100A

Specifications

About this item

Premium quality tubes provide superior protection for light sensitive applications.

Tubes are manufactured from amber 51 expansion borosilicate glass.

Packaging: Packed in convenient trays that are shrink-wrapped in plastic film.
Customers Who Bought This Also Bought