Crucibles Tall Form Glazed Porcelain, 30ml, 43x37x22mm, Tolerance +/-2.5%
Catalog #: CA10799-108
Supplier: SILICO & CHEMICO PORCELAIN SE
Crucibles Tall Form Glazed Porcelain, 30ml, 43x37x22mm, Tolerance +/-2.5%
Catalog #: CA10799-108
Supplier: SILICO & CHEMICO PORCELAIN SE
Customers Who Bought This Also Bought
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
$66.85 to $318.03
SP Bel-Art ProCulture Foil Cover-ALLS, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$86.77 to $708.50
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$464.49