Order Entry
ContactUsLinkComponent
Clamp
Premium Three-Finger Long Clamp with Bosshead, Vinyl Coated
Catalog #: 470165-412
Supplier:  Wards
Clamp
Premium Three-Finger Long Clamp with Bosshead, Vinyl Coated
Catalog #: 470165-412
Supplier:  Wards

Specifications

  • Description:
    Premium Three-Finger Long Clamp with Bosshead, Vinyl Coated
  • Cat. no.:
    470165-412
  • Supplier No.:
    PRCMP07BH

Specifications

About this item

Vinyl Lined Extension Lab Clamp

  • Maximum clamp opening is 8.5 cm.
  • Bosshead accommodates up to 19 mm diameter rod

Clamp constructed of powder coated steel, three rubber coated fingers attached to a high strength alloy swivel bosshead for ease of use in the lab.

Customers Who Bought This Also Bought