ContactUsLinkComponent
LanaGram
LanaGram™ Rapid Color Test, Hardy Diagnostics
Catalog #: CA10789-718
CAS Number:  
LanaGram
LanaGram™ Rapid Color Test, Hardy Diagnostics
Catalog #: CA10789-718
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    LanaGram, HDx
  • Size:
    15 tests per pack
  • Cat. no.:
    CA10789-718
Specifications
About this item
LanaGram™ rapid color test for bacterial gram reaction.
For organisms that are difficult to gram stain
LanaGram™ produces a rapid color change in a tube, gram negatives turn yellow, gram positives remain clear. Ideal for Bacillus, Listeria, Lactobacillus, and non-fermentors, all of which can be difficult to gram stain.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Uses the aminopeptidase reaction. LanaGram™ confirms the gram stain reaction of gram-negative and gram-positive aerobic or facultatively anaerobic rods or coccobacilli. This test is not recommended for use with gram-positive cocci, microaerophilic or obligate anaerobic organisms.
Customers Who Bought This Also Bought